Learn More
Ace Glass, Inc. -008 CHEMRAZ O-RING -008 CHEMRAZ O-RING
SDP
Catalog No. 509558103
Encompass
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Molded of perfluoroelastimer polymer, CHEMRAZ has the broadest chem- ical resistance of any elastomeric material. Combines the resiliance and sealing force of an elastomer with chemical resistance approaching that of PTFE. Resistant to swelling and embrittlement, easily in- stalled, conforming, not likely to creep or cold flow. O-ring,Chemraz,size 008
Catalog No. 50-955-8103 Supplier Ace Glass, Inc. Supplier No. 7859504
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Molded of perfluoroelastimer polymer, CHEMRAZ has the broadest chem- ical resistance of any elastomeric material. Combines the resiliance and sealing force of an elastomer with chemical resistance approaching that of PTFE. Resistant to swelling and embrittlement, easily in- stalled, conforming, not likely to creep or cold flow. O-ring,Chemraz,size 008