Learn More
Ace Glass, Inc. -008 O-RINGS VITON 12/PK -008 O-RINGS VITON 12/PK
SDP
Catalog No. 509558013
Encompass_Preferred
Size 008 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-955-8013 Supplier Ace Glass, Inc. Supplier No. 785504
Size 008 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.