Learn More
Ace Glass, Inc. -009 O-RING FETFE 12/PK -009 O-RING FETFE 12/PK
SDP
Catalog No. 509558045
Encompass_Preferred
Size 009 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-955-8045 Supplier Ace Glass, Inc. Supplier No. 7855707
Size 009 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.