Learn More
ACCUTEC BLADES INC (3P) .009" Stainless Steel 3-Facet Blades, Aluminum Back, Wrapped, 250 Blades per Carton, 20 Cartons per Case
Catalog No. NC3721099
Encompass_Preferred
AccuForge Single Edge Long blades feature a 2.25" long length for extra cutting width and 3-facet edges for increased sharpness. Our proprietary MicroCoat Enrichment Technology reduces cut force and helps maintain a sharp effective edge over multiple uses. Individually wrapped for safety. Also available in a smaller pack - see AGBL-7000-0000. Made in the USA.
Catalog No. NC3721099 Supplier ACCUTEC BLADES INC (3P) Supplier No. AGBL70010000
AccuForge Single Edge Long blades feature a 2.25" long length for extra cutting width and 3-facet edges for increased sharpness. Our proprietary MicroCoat Enrichment Technology reduces cut force and helps maintain a sharp effective edge over multiple uses. Individually wrapped for safety. Also available in a smaller pack - see AGBL-7000-0000. Made in the USA.