Learn More
ACCUTEC BLADES INC (3P) .009" Stainless Steel 3-Facet Degreased & Washed Blades, Stainless Steel Back, 100 Blades per Dispenser, 50 Dispensers per Case
Catalog No. NC3721079
Encompass_Preferred
AccuForge Pop-Up Dispenser provides quick, easy blade access for high volume applications. Blades feature .009" stainless steel and 3-Facet edges for sharpness. Stainless steel backing material provides optimal corrosion resistance. Blades are degreased and washed for least amount of residual oil. Each disposable dispenser contains 100 blades. Made in the USA
Catalog No. NC3721079 Supplier ACCUTEC BLADES INC (3P) Supplier No. AGBL70290000/2025
AccuForge Pop-Up Dispenser provides quick, easy blade access for high volume applications. Blades feature .009" stainless steel and 3-Facet edges for sharpness. Stainless steel backing material provides optimal corrosion resistance. Blades are degreased and washed for least amount of residual oil. Each disposable dispenser contains 100 blades. Made in the USA