Learn More
Pickering Laboratories Inc 012506H/PROTEIN HYDRO STD
SDP
Catalog No. NC9488696
Encompass_Preferred
Protein Hydrolysate Standard, in Sodium Citrate Buffer, 0.25 µmole/mL, each (5 mL/bottle) Amino Acid Analysis.
Catalog No. NC9488696 Supplier Pickering Laboratories Inc Supplier No. 012506H
Protein Hydrolysate Standard, in Sodium Citrate Buffer, 0.25 µmole/mL, each (5 mL/bottle) Amino Acid Analysis.