Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 0129 Discs

Catalog No. R65015 Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
R65015 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R65015 Supplier Thermo Scientific™ Supplier No. DD0015T
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate Vibrios from other Gram-negative rods using these diagnostic discs.

Differentiate vibrios from other Gram-negative rods using Thermo Scientific™ Oxoid™ 0129 Discs. The test exploits the sensitivity of vibrios to the vibrio static agent 0129 (2,4-Diamino-6,7-di-iso-propylpteridine phosphate)1. 0129 Discs are of great value in differentiating vibrios from other Gram-negative rods and particularly from aeromonads, which are characteristically resistant to 0129. The degree of sensitivity of vibrios to 0129 can also be used as a diagnostic feature in differentiation of Vibrio species.

50 Discs/Cartridge.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Concentration 150 μg
Detectable Analytes Vibrio species, Gram-negative rods
Format Disk, Cartridge
Type O129 Disc
Packaging Type 50 Discs/Cartridge
Quantity 150μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.