Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 15% w/w (L-106), Reagents

Catalog No. 01333585
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-585 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-585 Supplier Reagents Holdings Llc Supplier No. CS1526004A
Only null left
Add to Cart
Add to Cart

Sodium Chloride, 15% w/w (L-106), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Table salt solution
IUPAC Name water
SMILES O

Specifications

Boiling Point 103°C
Color Colorless
pH 5 to 9
Physical Form Liquid
Quantity 4 L
Percent Purity 15%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1105
Chemical Name or Material Sodium Chloride, 15% w/w (L-106)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.