Learn More
MSC Mitutoyo Dial Caliper, 150 mm, 0.02 mm Graduation, 1 ea
Catalog No. 502241271
Encompass
This dial caliper is designed for precise measurements in various applications. Constructed with hardened stainless steel, it offers durability and accuracy for outside, inside, depth, and step measurements.
  • Hardened stainless steel construction for extended life and corrosion resistance
  • Thumb roller for consistent feel and accurate measurements
  • Versatile for outside, inside, step, and depth measurements
  • TiN coated sliding surfaces on main beam to reduce wear
  • Extra rigid main beam for stability
  • Micro-fine graduations for precise readings
  • Adjustable and locking bezel
  • Covered rack keeps gear teeth clean
  • Easy zero adjustment
  • Satin chrome finish
  • Includes certificate of inspection
This item is not returnable. View return policy
Catalog No. 50-224-1271 Supplier MSC Supplier No. 84477702
This dial caliper is designed for precise measurements in various applications. Constructed with hardened stainless steel, it offers durability and accuracy for outside, inside, depth, and step measurements.
  • Hardened stainless steel construction for extended life and corrosion resistance
  • Thumb roller for consistent feel and accurate measurements
  • Versatile for outside, inside, step, and depth measurements
  • TiN coated sliding surfaces on main beam to reduce wear
  • Extra rigid main beam for stability
  • Micro-fine graduations for precise readings
  • Adjustable and locking bezel
  • Covered rack keeps gear teeth clean
  • Easy zero adjustment
  • Satin chrome finish
  • Includes certificate of inspection