Learn More
Bellco Glass, Inc. 0.2 mL PCR 8-strip Tubes, White, Sterile, 125/pk, 1250/cs
SDP
Catalog No. 502330652
Encompass_Preferred
0.1ml PCR 8-strip Tubes, Clear, Non-sterile, 125/pk, 1250/cs. has both clear and white 0.1/0.2 ml PCR tubes which respectively applies to the general PCR and qPCR reactions. Low profile PCR tubes can also reduce the evaporation of the reaction solution and increase the real-time PCR fluorescent signal strength. • Made from polypropylene• Compatible with most thermal cyclers and real-time PCR instruments• The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster• Easy to open leak proof cap makes the loss of reaction less than 0.2%.
Catalog No. 50-233-0652 Supplier Bellco Glass, Inc. Supplier No. 451400443
0.1ml PCR 8-strip Tubes, Clear, Non-sterile, 125/pk, 1250/cs. has both clear and white 0.1/0.2 ml PCR tubes which respectively applies to the general PCR and qPCR reactions. Low profile PCR tubes can also reduce the evaporation of the reaction solution and increase the real-time PCR fluorescent signal strength. • Made from polypropylene• Compatible with most thermal cyclers and real-time PCR instruments• The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster• Easy to open leak proof cap makes the loss of reaction less than 0.2%.