Learn More
Refrigiwear Inc. 0223R Regular Blue Large Polypro Liner
SDP
Catalog No. NC3867060
Encompass_Preferred
Glove liners provide an extra layer of protection against injury, help absorb perspiration, and increase the warmth of other gloves, or be worn alone when temperatures are moderate. The Polypropylene Glove Liner is moisture wicking and slides easily into other gloves when worn as a liner. Sold by the dozen, price shown is per pair. 100% Stretch polypropylene Moisture-wicking Sold by the dozen  
Catalog No. NC3867060 Supplier Refrigiwear Inc. Supplier No. 0223RBLULAR
Glove liners provide an extra layer of protection against injury, help absorb perspiration, and increase the warmth of other gloves, or be worn alone when temperatures are moderate. The Polypropylene Glove Liner is moisture wicking and slides easily into other gloves when worn as a liner. Sold by the dozen, price shown is per pair. 100% Stretch polypropylene Moisture-wicking Sold by the dozen  