Learn More
Refrigiwear Inc. 0231R Regular Black Large Merino Wool Liner
SDP
Catalog No. NC3867070
Encompass_Preferred
No other natural material can compete with wool for warmness and comfort. The Stretch Wool Glove Liner is made with 91% Merino wool and 9% synthetic fibers. The liner fits most sizes and easily stretches to maximize comfort. Sold by the dozen, price shown is per pair. To clean your wool items, hand wash in warm water with a mild detergent. Dry them by laying them flat on a surface. Do not hang dry as this may distort their shape. 91% Merino wool and 9% spandex Stretch-fit for maximum comfort Sold by the dozen    
Catalog No. NC3867070 Supplier Refrigiwear Inc. Supplier No. 0231RBLKLAR
No other natural material can compete with wool for warmness and comfort. The Stretch Wool Glove Liner is made with 91% Merino wool and 9% synthetic fibers. The liner fits most sizes and easily stretches to maximize comfort. Sold by the dozen, price shown is per pair. To clean your wool items, hand wash in warm water with a mild detergent. Dry them by laying them flat on a surface. Do not hang dry as this may distort their shape. 91% Merino wool and 9% spandex Stretch-fit for maximum comfort Sold by the dozen    