Learn More
Perkin Elmer US LLC 0.25mm i.d. Connecting Line (green), DXi Version
Catalog No. 502112128
Encompass_Preferred
0.25mm i.d. (green) Connects the FAST valve to an ESI ST nebulizer to achieve fast signal stabilization time and short wash times. Attaches to port #3 of the FAST valve. (DXi Version)
Catalog No. 50-211-2128 Supplier Perkin Elmer US LLC Supplier No. N0777655
0.25mm i.d. (green) Connects the FAST valve to an ESI ST nebulizer to achieve fast signal stabilization time and short wash times. Attaches to port #3 of the FAST valve. (DXi Version)