Learn More
Bellco Glass, Inc. 0.2ml 96 Well PCR Plate, No Skirt, Clear, H1 notch, 25/pk, 100/cs
SDP
Catalog No. 502329259
Encompass_Preferred
0.2ml 96 Well PCR Plate, No Skirt, Clear, H1 notch, 25/pk, 100/cs. When using a thermal cycler with fluorescence detector above the samples, white PCR plates effectively prevent crosstalk between wells. The white PCR plate also allows for high accuracy of detection when signal intensity is low. clear PCR plates are designed for PCR machines where the fluorescence detector is located below the sample plate. • Made of high quality polypropylene to ensure minimal loss of reaction solution.• Flat surface, thick and solid, not easy to deform.• Elevated edge of the hole can better prevent cross contamination.• Compatible with pressure sensitive film, self-adhesive film and hot sealing film.• Autoclavable.
Catalog No. 50-232-9259 Supplier Bellco Glass, Inc. Supplier No. 451400454
0.2ml 96 Well PCR Plate, No Skirt, Clear, H1 notch, 25/pk, 100/cs. When using a thermal cycler with fluorescence detector above the samples, white PCR plates effectively prevent crosstalk between wells. The white PCR plate also allows for high accuracy of detection when signal intensity is low. clear PCR plates are designed for PCR machines where the fluorescence detector is located below the sample plate. • Made of high quality polypropylene to ensure minimal loss of reaction solution.• Flat surface, thick and solid, not easy to deform.• Elevated edge of the hole can better prevent cross contamination.• Compatible with pressure sensitive film, self-adhesive film and hot sealing film.• Autoclavable.