Learn More
Refrigiwear Inc. 0353R Regular White XLarge Premium Insulated Goatskin
SDP
Catalog No. NC3867113
Encompass_Preferred
Smash through your to-do list, not your fingers. The Ergo Goatskin Glove features flexible goat grain leather armored with impact protection pads on the fingers and across the knuckles. These leather work gloves also deliver triple-layer warmth against temperatures as low as -25�F with fiberfill and foam insulation and a soft tricot lining to protect against the harshest conditions. These insulated work gloves also come with a pre-curved ergonomic design and a durable reinforced palm to prevent hand fatigue. When the conditions are tough, you need tougher gloves. Reach for the Ergo Goatskin Glove and get it done. Comfort Rating: -25�F Fiberfill and foam insulation with tricot lining Goat grain and spandex outershell Pre-curved, ergonomic fit Reinforced thumb crotch Impact protection on fingers, knuckles and back Rolled Fingertips Stretch neoprene cuff  
Catalog No. NC3867113 Supplier Refrigiwear Inc. Supplier No. 0353RWHTXLG
Smash through your to-do list, not your fingers. The Ergo Goatskin Glove features flexible goat grain leather armored with impact protection pads on the fingers and across the knuckles. These leather work gloves also deliver triple-layer warmth against temperatures as low as -25�F with fiberfill and foam insulation and a soft tricot lining to protect against the harshest conditions. These insulated work gloves also come with a pre-curved ergonomic design and a durable reinforced palm to prevent hand fatigue. When the conditions are tough, you need tougher gloves. Reach for the Ergo Goatskin Glove and get it done. Comfort Rating: -25�F Fiberfill and foam insulation with tricot lining Goat grain and spandex outershell Pre-curved, ergonomic fit Reinforced thumb crotch Impact protection on fingers, knuckles and back Rolled Fingertips Stretch neoprene cuff  