Learn More
Refrigiwear Inc. 0407R Regular Black/Blue Lar ProWeight Thermal ErgoGrip
SDP
Catalog No. NC3867117
Encompass_Preferred
The ProWeight Thermal Ergo Glove is a tear-resistant glove with a proweight terry lining, an ergonomic fit and a textured waterproof palm to give you extra grip and protection. 7-gauge brushed acrylic shell Pro-weight terry lining Crinkle finish latex palm coating Pre-curved, ergonomic fit  
Catalog No. NC3867117 Supplier Refrigiwear Inc. Supplier No. 0407RBBKLAR
The ProWeight Thermal Ergo Glove is a tear-resistant glove with a proweight terry lining, an ergonomic fit and a textured waterproof palm to give you extra grip and protection. 7-gauge brushed acrylic shell Pro-weight terry lining Crinkle finish latex palm coating Pre-curved, ergonomic fit  