Learn More
Refrigiwear Inc. 0408R Regular HiVis Lime XLG HiVis Doubl ProWeight Ergogrip
SDP
Catalog No. NC3867136
Encompass_Preferred
The Dual-Layer HiVis Ergo Glove features a tear-resistant, terry lining with an ergonomic fit and textured waterproof palm to give you extra grip and protection. HiVis color adds extra visibility in low-light conditions. Dual Layer: Outershell and built-in liner for extra warmth 15-Gauge polyester shell with foam nitrile smooth finish coating and double nitrile sandy finish palm coating 7-Gauge heavy brushed acrylic liner Pre-curved ergonomic fit  
Catalog No. NC3867136 Supplier Refrigiwear Inc. Supplier No. 0408RHVLXLG
The Dual-Layer HiVis Ergo Glove features a tear-resistant, terry lining with an ergonomic fit and textured waterproof palm to give you extra grip and protection. HiVis color adds extra visibility in low-light conditions. Dual Layer: Outershell and built-in liner for extra warmth 15-Gauge polyester shell with foam nitrile smooth finish coating and double nitrile sandy finish palm coating 7-Gauge heavy brushed acrylic liner Pre-curved ergonomic fit  