Learn More
Braintree Scientific .040IN O.D> X .025IN I.D. 50FT
SDP
Catalog No. NC9053366
Encompass
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**
Catalog No. NC9053366 Supplier Braintree Scientific Supplier No. MRE04050FT
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**