Learn More
Refrigiwear Inc. 0419R Regular Gold Large Double Insulated Cowhide
SDP
Catalog No. NC3867140
Encompass_Preferred
The Double Insulated Cowhide Leather Glove keeps you going in extreme cold with double the insulation and a rugged split cowhide exterior. Abrasion pads on fingers and palm on this leather freezer glove give it extra durability, so it's up for the toughest jobs. Please note that leather is not treated at the factory for waterproofing. Comfort Rating: -30�F Fiberfill plus, thick double-foam insulation with tricot lining Split cowhide leather PVC abrasion pads on fingers and palm Safety cuff with heavy-duty knit wrist Kevlar® thread Improved fit Meets EN 511:2006 Protective Gloves Against Cold: 3 | 4 | 0 Meets EN 388:2016 Protective Gloves Against Mechanical Risks: 4 | 2 | 4 | 4 | X  
Catalog No. NC3867140 Supplier Refrigiwear Inc. Supplier No. 0419RGLDLAR
The Double Insulated Cowhide Leather Glove keeps you going in extreme cold with double the insulation and a rugged split cowhide exterior. Abrasion pads on fingers and palm on this leather freezer glove give it extra durability, so it's up for the toughest jobs. Please note that leather is not treated at the factory for waterproofing. Comfort Rating: -30�F Fiberfill plus, thick double-foam insulation with tricot lining Split cowhide leather PVC abrasion pads on fingers and palm Safety cuff with heavy-duty knit wrist Kevlar® thread Improved fit Meets EN 511:2006 Protective Gloves Against Cold: 3 | 4 | 0 Meets EN 388:2016 Protective Gloves Against Mechanical Risks: 4 | 2 | 4 | 4 | X  