Learn More
Refrigiwear Inc. 0421R REGULAR GREEN XLARGE Insulated Wool Silicone Grip
SDP
Catalog No. NC3885942
Encompass_Preferred
This Insulated Wool Grip Glove is made of 100% Ragg wool, which provides natural insulation and wicking, with insulation and a fleece lining for additional warmth and comfort. Herringbone style silicone grip performs better in the cold, without cracking or getting slick. To clean your wool items, hand wash in warm water with a mild detergent. Leave in the water for about ten minutes and dry them by laying them flat on a surface. Do not hang dry as this may distort their shape. Comfort Rating: 10°F 40g Insulation plus fleece lining Ragg wool outershell Silicone herringbone grip on palm Extended double layer knit cuff    
Catalog No. NC3885942 Supplier Refrigiwear Inc. Supplier No. 0421RGRNXLG
This Insulated Wool Grip Glove is made of 100% Ragg wool, which provides natural insulation and wicking, with insulation and a fleece lining for additional warmth and comfort. Herringbone style silicone grip performs better in the cold, without cracking or getting slick. To clean your wool items, hand wash in warm water with a mild detergent. Leave in the water for about ten minutes and dry them by laying them flat on a surface. Do not hang dry as this may distort their shape. Comfort Rating: 10°F 40g Insulation plus fleece lining Ragg wool outershell Silicone herringbone grip on palm Extended double layer knit cuff    