Learn More
Perkin Elmer US LLC 0.5 µL Low Injection Volume Syringe, Metal Plunger 0.63 mm o.d. needle BLUE Barrel (pack of 10 syringes)
Catalog No. 502111530
Encompass_Preferred
0.5 µL Low Injection Volume Syringe, Metal Plunger 0.63 mm o.d. needle BLUE Barrel (pack of 10 syringes)
Catalog No. 50-211-1530 Supplier Perkin Elmer US LLC Supplier No. N6103243
0.5 µL Low Injection Volume Syringe, Metal Plunger 0.63 mm o.d. needle BLUE Barrel (pack of 10 syringes)