Learn More
Perkin Elmer US LLC 0.5 mm I.D. Internal Standards Probe for NexION 2000/1000
Catalog No. 502112194
Encompass_Preferred
The Internal Standards Probes offer high chemical resistance for applications involving strong acids, alkalis and organics. A contoured design reduces carryover, while an all-PFA-wetted sample flow path reduces contamination. The probes can be used with the MEINHARD Nebulizer and the PFA Nebulizer. The probe is included in the Organics Sample Introduction Kit (N8150031), the NexION 2000 ICP-MS Base Configuration's Sample Introduction Kit (N8150034), the Core Configuration's Sample Introduction Kit (N8150035), and the Semi Configuration's Sample Introduction Kit (N8150037).
Catalog No. 50-211-2194 Supplier Perkin Elmer US LLC Supplier No. N8152452
The Internal Standards Probes offer high chemical resistance for applications involving strong acids, alkalis and organics. A contoured design reduces carryover, while an all-PFA-wetted sample flow path reduces contamination. The probes can be used with the MEINHARD Nebulizer and the PFA Nebulizer. The probe is included in the Organics Sample Introduction Kit (N8150031), the NexION 2000 ICP-MS Base Configuration's Sample Introduction Kit (N8150034), the Core Configuration's Sample Introduction Kit (N8150035), and the Semi Configuration's Sample Introduction Kit (N8150037).