Learn More
Refrigiwear Inc. 0509R Regular Black Large Dbl Layr Herringbne Grip Glove
SDP
Catalog No. NC3867095
Encompass_Preferred
Warm and functional, the Dual Layer Herringbone Glove has a dual-layer with built-in lining for added warmth and protection, so no liner is needed. The herringbone style silicone grip performs better in cold, without cracking or getting slick. Dual Layer: Outershell and built-in liner for extra warmth Acrylic outershell with brushed acrylic loop terry liner Silicone herringbone palm coating  
Catalog No. NC3867095 Supplier Refrigiwear Inc. Supplier No. 0509RBLKLAR
Warm and functional, the Dual Layer Herringbone Glove has a dual-layer with built-in lining for added warmth and protection, so no liner is needed. The herringbone style silicone grip performs better in cold, without cracking or getting slick. Dual Layer: Outershell and built-in liner for extra warmth Acrylic outershell with brushed acrylic loop terry liner Silicone herringbone palm coating  