Learn More
Teknova 0.5M MES Buffer, pH 6.1. 500mL, Sterile.
SDP
Catalog No. 502388025
Encompass
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.
Catalog No. 50-238-8025 Supplier Teknova Supplier No. M2225
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.