Learn More
Teknova 0.5M Tris, 1.5M Sodium Chloride, 0.5% Tween-20, pH 7.7. 500mL, Sterile.
SDP
Catalog No. 502388180
Encompass
0.5M Tris,1.5M Sodium Chloride,0.5% Tween-20
Catalog No. 50-238-8180 Supplier Teknova Supplier No. 2A7665
0.5M Tris,1.5M Sodium Chloride,0.5% Tween-20