Learn More
Perkin Elmer US LLC 0.5mm i.d. (orange) FAST Connector for Gem Cone Nebulizer
Catalog No. 502112155
Encompass_Preferred
0.5mm i.d. (orange) FAST connector for PerkinElmer Gem Cone Nebulizer. Attaches to port #3 of the FAST valve.
Catalog No. 50-211-2155 Supplier Perkin Elmer US LLC Supplier No. N0777659
0.5mm i.d. (orange) FAST connector for PerkinElmer Gem Cone Nebulizer. Attaches to port #3 of the FAST valve.