Learn More
Perkin Elmer US LLC 0.5mm i.d. (orange) FAST connector for Miramist Nebulizer
Catalog No. 502112125
Encompass_Preferred
0.5mm i.d. (orange) FAST connector for Miramist Nebulizer. Attaches to port #3 of the FAST valve.
Catalog No. 50-211-2125 Supplier Perkin Elmer US LLC Supplier No. N0777658
0.5mm i.d. (orange) FAST connector for Miramist Nebulizer. Attaches to port #3 of the FAST valve.