Learn More
Avanti Polar Lipids 1,2-dihexanoyl-sn-glycero-3-phosphocholine, 34506-67-7, MFCD00050431, 25 mg
Catalog No. NC1453363
Encompass
Molecular Formula: C20H40NO8P, Formula Weight: 453.507, Purity >99%.
Catalog No. NC1453363 Supplier Avanti Polar Lipids Supplier No. 850305C25MG
Molecular Formula: C20H40NO8P, Formula Weight: 453.507, Purity >99%.