Learn More
MSC Mitutoyo Dial Caliper, 6 in, 0.001 Grad, Stainless, 1 ea
Catalog No. 502241184
Encompass
This dial caliper features hardened stainless steel construction, a thumb roller for consistent measurements, and is designed for outside, inside, step, and depth measurements. Its main beam's sliding surfaces are Titanium Nitride (TiN) coated to reduce wear, ensuring durability and precise operation.
  • Hardened stainless steel construction for durability and corrosion resistance
  • Thumb roller for consistent and accurate measurements
  • TiN coated main beam sliding surfaces to reduce wear
  • Extra rigid main beam
  • Micro-fine graduations
  • Adjustable and locking bezel
  • Covered rack to keep gear teeth clean
  • Easy zero adjustment
  • Satin chrome finish
  • Supplied in a fitted plastic case
  • Performs outside, inside, step, and depth measurements
This item is not returnable. View return policy
Catalog No. 50-224-1184 Supplier MSC Supplier No. 84477777
This dial caliper features hardened stainless steel construction, a thumb roller for consistent measurements, and is designed for outside, inside, step, and depth measurements. Its main beam's sliding surfaces are Titanium Nitride (TiN) coated to reduce wear, ensuring durability and precise operation.
  • Hardened stainless steel construction for durability and corrosion resistance
  • Thumb roller for consistent and accurate measurements
  • TiN coated main beam sliding surfaces to reduce wear
  • Extra rigid main beam
  • Micro-fine graduations
  • Adjustable and locking bezel
  • Covered rack to keep gear teeth clean
  • Easy zero adjustment
  • Satin chrome finish
  • Supplied in a fitted plastic case
  • Performs outside, inside, step, and depth measurements