Learn More
ADMINMED NANOBIOSCIENCES 0600UM MICRONEEDLE ARRAY
SDP
Catalog No. NC0584391
Encompass
AdminPatch Array 600 is a microneedle array, which has 187 (one hundred eighty seven) 500 um-tall microneedles located within 1 sq. cm circular area. The entire device is 20 mm in diameter and is made of medical-grade SS316L stainless steel.
Catalog No. NC0584391 Supplier ADMINMED NANOBIOSCIENCES Supplier No. SKU104
AdminPatch Array 600 is a microneedle array, which has 187 (one hundred eighty seven) 500 um-tall microneedles located within 1 sq. cm circular area. The entire device is 20 mm in diameter and is made of medical-grade SS316L stainless steel.