Learn More
Bellco Glass, Inc. 0.6ml Microcentrifuge Tube, Clear, Conical, Lock Cap,1000/pk, 10000/cs
SDP
Catalog No. 502330721
Encompass_Preferred
0.6ml Microcentrifuge Tube, Clear, Conical, Lock Cap,1000/pk, 10000/cs. centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagents. • USP VI grade polypropolyne material, free from heavy metal• RCF: 30000 x g• Endotoxin level: <0.1EU• Clear graduations and frosted marking area• Special safe lock cap design provides secure lid sealing• Easy opening and closing• Temperature range: -80℃ to +121℃• DNase/RNase free• Tubes are autoclavable at 121°C (250°F), under 15psi for 10–15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity. • Also available as pre-sterilized
Catalog No. 50-233-0721 Supplier Bellco Glass, Inc. Supplier No. 451400288
Add to Cart
Add to Cart
0.6ml Microcentrifuge Tube, Clear, Conical, Lock Cap,1000/pk, 10000/cs. centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagents. • USP VI grade polypropolyne material, free from heavy metal• RCF: 30000 x g• Endotoxin level: <0.1EU• Clear graduations and frosted marking area• Special safe lock cap design provides secure lid sealing• Easy opening and closing• Temperature range: -80℃ to +121℃• DNase/RNase free• Tubes are autoclavable at 121°C (250°F), under 15psi for 10–15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity. • Also available as pre-sterilized