Learn More
AVANTOR MASTERFLEX (VWR LLC) Standard Pump Head for Precision Tubing L/S 16, Polycarbonate Housing, SS Rotor
Catalog No. NC1827103
Encompass
Masterflex L/S Standard Pump Head for Precision Tubing L/S 16, Polycarbonate Housing, SS Rotor,Ideal for fluid transfer applications requiring high accuracy and repeatabilityStandard heads are precision molded to accommodate a single size of tubingPump head is compact and simple in design with same side tubing entry/exit¿perfect for space-limited applicationsStandard heads are compact and easy to integrate into your OEM applicationsStack up to four heads depending on drive recommendations
Catalog No. NC1827103 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0701621
Masterflex L/S Standard Pump Head for Precision Tubing L/S 16, Polycarbonate Housing, SS Rotor,Ideal for fluid transfer applications requiring high accuracy and repeatabilityStandard heads are precision molded to accommodate a single size of tubingPump head is compact and simple in design with same side tubing entry/exit¿perfect for space-limited applicationsStandard heads are compact and easy to integrate into your OEM applicationsStack up to four heads depending on drive recommendations