Learn More
Perkin Elmer US LLC Brownlee Aquapore ODS RP-18 NewGuard Cartridge - 15 mm x 3.2 mm I.D., Pkg. 3
Catalog No. NC9240922
Encompass_Preferred
Aquapore ODS RP-18 NewGuard cartridges are based on a spherical silica support suitable for the separation of large biopolymers. These cartridges prolong column life by 2 to 5 times while protecting columns from particulates and strongly bound sample components. Actual bed length of each 15 mm NewGuard is approximately 13 mm. These NewGuard Cartridges require a holder: To couple with a conventional column - NewGuard Holder (07150001) To couple with an MPLC cartridge - Union (07150018) Quantity: Package of 3 Interaction Mode: Reversed Phase USP Code: L1
Catalog No. NC9240922 Supplier Perkin Elmer US LLC Supplier No. 07110092
Aquapore ODS RP-18 NewGuard cartridges are based on a spherical silica support suitable for the separation of large biopolymers. These cartridges prolong column life by 2 to 5 times while protecting columns from particulates and strongly bound sample components. Actual bed length of each 15 mm NewGuard is approximately 13 mm. These NewGuard Cartridges require a holder: To couple with a conventional column - NewGuard Holder (07150001) To couple with an MPLC cartridge - Union (07150018) Quantity: Package of 3 Interaction Mode: Reversed Phase USP Code: L1