Learn More
Medequip Depot U-100 Syringe, 29G x 1/2 in. , 3/10cc, 100/box
SDP
Catalog No. NC1744254
Encompass
UltiMed, Inc. 09230 U-100 Syringe, 29G x 1/2 in. , 3/10cc, 100/bx. Precision engineered to provide superior comfort. Features include a triple beveled tip for easier insertion, electromagnetically polished for a smoother finish and silicone coated for added glide. Latex-free.
Catalog No. NC1744254 Supplier Medequip Depot Supplier No. 09230
UltiMed, Inc. 09230 U-100 Syringe, 29G x 1/2 in. , 3/10cc, 100/bx. Precision engineered to provide superior comfort. Features include a triple beveled tip for easier insertion, electromagnetically polished for a smoother finish and silicone coated for added glide. Latex-free.