Learn More
Edge Biosystems Inc 0ERFORMA SPIN COLUMN
SDP
Catalog No. NC0609033
Encompass
Performa� Spin Columns feature a fully hydrated gel matrix that provides for a convenient and simple method to purify your DNA/RNA samples.They effectively remove salts, amino acids, nucleotides, traces of solvents and other low molecular weight materials from nucleic acid fragments greater than 20 bases.
Catalog No. NC0609033 Supplier Edge Biosystems Inc Supplier No. 13266
Performa� Spin Columns feature a fully hydrated gel matrix that provides for a convenient and simple method to purify your DNA/RNA samples.They effectively remove salts, amino acids, nucleotides, traces of solvents and other low molecular weight materials from nucleic acid fragments greater than 20 bases.