Learn More
Leica Biosystems 0LIDE BOND ADHESIVE 1440/CS
Catalog No. NC0849950
Encompass_Preferred
Validated* slide for the BOND platformThe Apex BOND is an extension of the Apex Adhesive Slides portfolio that is specifically designed and validated for IHC staining on the BOND platform. The eight dots represent the usable areas of the Apex BOND for the different dispense volumes in the BOND processing modules.
Catalog No. NC0849950 Supplier Leica Biosystems Supplier No. 3800040
Validated* slide for the BOND platformThe Apex BOND is an extension of the Apex Adhesive Slides portfolio that is specifically designed and validated for IHC staining on the BOND platform. The eight dots represent the usable areas of the Apex BOND for the different dispense volumes in the BOND processing modules.