Learn More
Alkali Scientific 0.1ml Biorad Type 96 Well PCR Plate, No Skirt, White, 25/pk, 100/cs
SDP
Catalog No. 502336012
Encompass_Preferred
* Made from polypropylene* Compatible with most thermal cyclers and real-time PCR instruments* The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster* Easy to open leak proof cap makes the loss of reaction less than 0.2%
Catalog No. 50-233-6012 Supplier Alkali Scientific Supplier No. PCZ96
* Made from polypropylene* Compatible with most thermal cyclers and real-time PCR instruments* The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster* Easy to open leak proof cap makes the loss of reaction less than 0.2%