Learn More
Alkali Scientific 0.5mL, Microtube with Cap, Assorted Colors (Blue, Red, Green, Orange, Purple, Yellow), with Self-standing Bag, 500/cs
SDP
Catalog No. 502336088
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 0.5mL centrifuge tubes in assorted colors.
Catalog No. 50-233-6088 Supplier Alkali Scientific Supplier No. C3004AST
Add to Cart
Add to Cart
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 0.5mL centrifuge tubes in assorted colors.