Learn More
Alkali Scientific 0.5mL Qwik-Snap™ Clear Microtube with Cap, Sterile, in Self-standing Bag, 500/cs
SDP
Catalog No. 502336027
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. 500 clear, sterile 0.5mL centrifuge tubes per pack.
Catalog No. 50-233-6027 Supplier Alkali Scientific Supplier No. C3004S
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. 500 clear, sterile 0.5mL centrifuge tubes per pack.