Learn More
Alkali Scientific Qwik-Snap™ 0.6mL MicroTubes, Sterile, 50/Bag, 7500/cs
SDP
Catalog No. 502336229
Encompass_Preferred
Alkali Scientific Qwik-Snap™ 0.6mL MicroTubes are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. Certified RNase, DNase and pyrogen free. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80° C, the Qwik-Snapª micro centrifuge tubes are the perfect choice for your research.
Catalog No. 50-233-6229 Supplier Alkali Scientific Supplier No. C3009S
Alkali Scientific Qwik-Snap™ 0.6mL MicroTubes are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. Certified RNase, DNase and pyrogen free. Capable of withstanding boiling temperatures up to 121° C and freezing down to -80° C, the Qwik-Snapª micro centrifuge tubes are the perfect choice for your research.