Learn More
Alkali Scientific ClearSeal™ 0.5ml Sterile Screw Caps, Free Standing, Sterile, 5 Inner Packs of 100, 500/cs
SDP
Catalog No. 502335817
Encompass_Preferred
ClearSeal™ 0.5ml Sterile Screw Caps are made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal™ screw cap microtubes are sterile, RNase, DNase, and pyrogen-free and may be centrifuged at up to 22,000xg*.
Catalog No. 50-233-5817 Supplier Alkali Scientific Supplier No. C4805
ClearSeal™ 0.5ml Sterile Screw Caps are made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal™ screw cap microtubes are sterile, RNase, DNase, and pyrogen-free and may be centrifuged at up to 22,000xg*.