Learn More
Perkin Elmer US LLC 0-Slot Demountable Quartz Torch for Optima 8x00
Catalog No. 502112268
Encompass_Preferred
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for organic solvents.
Catalog No. 50-211-2268 Supplier Perkin Elmer US LLC Supplier No. N0780133
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for organic solvents.