Learn More
Rockland Immunochemicals 1.0 M Tris HCl pH 7.4 - MB-002
SDP
Catalog No. NC1283464
Encompass_Preferred
Tris is extensively used in biochemistry and molecular biology. In biochemistry, Tris is widely used as a component of additional buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids. Tris HCl solution (tris(hydroxymethyl)aminomethane hydrochloride) is useful for stabilization of buffer formulations and electrophoresis of biological molecules. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers.
Catalog No. NC1283464 Supplier Rockland Immunochemicals Supplier No. MB002
Tris is extensively used in biochemistry and molecular biology. In biochemistry, Tris is widely used as a component of additional buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids. Tris HCl solution (tris(hydroxymethyl)aminomethane hydrochloride) is useful for stabilization of buffer formulations and electrophoresis of biological molecules. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers.