Learn More
Rockland Immunochemicals 1.0 M Tris HCl pH 7.6 - MB-003
SDP
Catalog No. 501058965
Encompass_Preferred
Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology. In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
Catalog No. 50-105-8965 Supplier Rockland Immunochemicals Supplier No. MB003
Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology. In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.