Learn More
Rockland Immunochemicals 1.0 M Tris HCl pH 9.0 - MB-028-1000
SDP
Catalog No. 501058991
Encompass_Preferred
Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology. In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
Catalog No. 50-105-8991 Supplier Rockland Immunochemicals Supplier No. MB0281000
Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology. In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.