Learn More
Tisch Scientific Nylon Syringe Filters, 1.0um, 25mm, Luer-Lok/Luer Slip, Nonsterile, 100/Pk
SDP
Catalog No. NC1982755
Encompass
Featuring naturally hydrophilic nylon membrane technology with 1.0µm pore structure engineered for efficient coarse filtration and particulate removal applications. The SF17041 delivers reliable 100ml/minute flow rates through 4.08cm² active membrane area, optimizing throughput for routine sample clarification procedures. Tisch Scientific syringe filters incorporate durable polypropylene housing with integrated prefilter technology, engineered to withstand 87psi operating pressure during demanding laboratory protocols.
Catalog No. NC1982755 Supplier Tisch Scientific Supplier No. SF17041
Featuring naturally hydrophilic nylon membrane technology with 1.0µm pore structure engineered for efficient coarse filtration and particulate removal applications. The SF17041 delivers reliable 100ml/minute flow rates through 4.08cm² active membrane area, optimizing throughput for routine sample clarification procedures. Tisch Scientific syringe filters incorporate durable polypropylene housing with integrated prefilter technology, engineered to withstand 87psi operating pressure during demanding laboratory protocols.