Learn More
ERA WATERS CORP 1.00MG/L C NIST KHP-40ML
SDP
Catalog No. NC1862944
Encompass
"1.00 mg/L C NIST KHP, 40 mL NIST KHP CRMs.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."
Catalog No. NC1862944 Supplier ERA WATERS CORP Supplier No. 11800ERA
"1.00 mg/L C NIST KHP, 40 mL NIST KHP CRMs.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."