Learn More
Rockland Immunochemicals 1.0 M HEPES pH 7.2 - MB-062-0100
SDP
Catalog No. NC0746234
Encompass_Preferred
HEPES is a useful buffer for pH's ranging from 6.8-8.2. This makes HEPES ideal for cell culturing because of its usefulness in maintaining pH changes in presence of carbon dioxide production from cellular respiration.1.0 M HEPES pH 7.2 is ideal for investigators in Molecular Biology, Cell Biology, and Immunology researchers.
Catalog No. NC0746234 Supplier Rockland Immunochemicals Supplier No. MB0620100
HEPES is a useful buffer for pH's ranging from 6.8-8.2. This makes HEPES ideal for cell culturing because of its usefulness in maintaining pH changes in presence of carbon dioxide production from cellular respiration.1.0 M HEPES pH 7.2 is ideal for investigators in Molecular Biology, Cell Biology, and Immunology researchers.