Learn More
Thomas Scientific 1.0ml 2D Barcoded ScrewTop Tubes precapped with Orange Internal Screwcap bulk, 960 pack
SDP
Catalog No. NC1758211
Encompass
2D-barcode laser etched in high-contrast on base of FluidX sample storage tubes offers a secure audit trail for sample identification and sample tracking, and a permanent link between sample and data.
Catalog No. NC1758211 Supplier Thomas Scientific Supplier No. 1150N59
2D-barcode laser etched in high-contrast on base of FluidX sample storage tubes offers a secure audit trail for sample identification and sample tracking, and a permanent link between sample and data.