Learn More
Analytical Sales & Services 1.0mL 96-Well Opaque Flexi-Tier™ Block System
Catalog No. 5000807180
Encompass
1.0mL 96-Well Opaque Flexi-Tier System\n\nIncludes:\n• 1.0mL Conical Glass Inserts\n• Spacer\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH
Catalog No. 50-008-07180 Supplier Analytical Sales & Services Supplier No. 96FP10
Add to Cart
Add to Cart
1.0mL 96-Well Opaque Flexi-Tier System\n\nIncludes:\n• 1.0mL Conical Glass Inserts\n• Spacer\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH